MilliporeSigma
An immunogenic personal neoantigen vaccine for patients with melanoma.
Nature (2017-07-06)
Patrick A Ott, Zhuting Hu, Derin B Keskin, Sachet A Shukla, Jing Sun, David J Bozym, Wandi Zhang, Adrienne Luoma, Anita Giobbie-Hurder, Lauren Peter, Christina Chen, Oriol Olive, Todd A Carter, Shuqiang Li, David J Lieb, Thomas Eisenhaure, Evisa Gjini, Jonathan Stevens, William J Lane, Indu Javeri, Kaliappanadar Nellaiappan, Andres M Salazar, Heather Daley, Michael Seaman, Elizabeth I Buchbinder, Charles H Yoon, Maegan Harden, Niall Lennon, Stacey Gabriel, Scott J Rodig, Dan H Barouch, Jon C Aster, Gad Getz, Kai Wucherpfennig, Donna Neuberg, Jerome Ritz, Eric S Lander, Edward F Fritsch, Nir Hacohen, Catherine J Wu
ABSTRACT
Effective anti-tumour immunity in humans has been associated with the presence of T cells directed at cancer neoantigens, a class of HLA-bound peptides that arise from tumour-specific mutations. They are highly immunogenic because they are not present in normal tissues and hence bypass central thymic tolerance. Although neoantigens were long-envisioned as optimal targets for an anti-tumour immune response, their systematic discovery and evaluation only became feasible with the recent availability of massively parallel sequencing for detection of all coding mutations within tumours, and of machine learning approaches to reliably predict those mutated peptides with high-affinity binding of autologous human leukocyte antigen (HLA) molecules. We hypothesized that vaccination with neoantigens can both expand pre-existing neoantigen-specific T-cell populations and induce a broader repertoire of new T-cell specificities in cancer patients, tipping the intra-tumoural balance in favour of enhanced tumour control. Here we demonstrate the feasibility, safety, and immunogenicity of a vaccine that targets up to 20 predicted personal tumour neoantigens. Vaccine-induced polyfunctional CD4
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 18-39 human, ≥97% (HPLC)