MilliporeSigma
Lamtor1 Is Critically Required for CD4
Journal of immunology (Baltimore, Md. : 1950) (2017-08-05)
Takashi Hosokawa, Tetsuya Kimura, Shigeyuki Nada, Tatsusada Okuno, Daisuke Ito, Sujin Kang, Satoshi Nojima, Kazuya Yamashita, Takeshi Nakatani, Yoshitomo Hayama, Yasuhiro Kato, Yuhei Kinehara, Masayuki Nishide, Norihisa Mikami, Syohei Koyama, Hyota Takamatsu, Daisuke Okuzaki, Naganari Ohkura, Shimon Sakaguchi, Masato Okada, Atsushi Kumanogoh
ABSTRACT
Mechanistic target of rapamycin complex (mTORC)1 integrates intracellular sufficiency of nutrients and regulates various cellular functions. Previous studies using mice with conditional knockout of mTORC1 component proteins (i.e., mTOR, Raptor, and Rheb) gave conflicting results on the roles of mTORC1 in CD4
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Pertussis toxin from Bordetella pertussis, lyophilized powder