MilliporeSigma
  • Activation of Sirt1/FXR Signaling Pathway Attenuates Triptolide-Induced Hepatotoxicity in Rats.
Activation of Sirt1/FXR Signaling Pathway Attenuates Triptolide-Induced Hepatotoxicity in Rats.
Frontiers in pharmacology (2017-05-26)
Jing Yang, Lixin Sun, Lu Wang, Hozeifa M Hassan, Xuan Wang, Phillip B Hylemon, Tao Wang, Huiping Zhou, Luyong Zhang, Zhenzhou Jiang
ABSTRACT
Triptolide (TP), a diterpenoid isolated from
Sigma-Aldrich
GW4064, ≥97% (HPLC)