MilliporeSigma
  • Brain Pharmacokinetics and the Pharmacological Effects on Striatal Neurotransmitter Levels of
Brain Pharmacokinetics and the Pharmacological Effects on Striatal Neurotransmitter Levels of
Frontiers in pharmacology (2017-09-21)
Bingxin Xiao, Zengxian Sun, Fangrui Cao, Lisha Wang, Yonghong Liao, Xinmin Liu, Ruile Pan, Qi Chang
ABSTRACT
Isoflavonoids are putatively active components of
Sigma-Aldrich
2-(aminomethyl)phenol, AldrichCPR
Supelco
Serotonin, analytical standard