MilliporeSigma
  • Ultrathin Nafion-filled porous membrane for zinc/bromine redox flow batteries.
Ultrathin Nafion-filled porous membrane for zinc/bromine redox flow batteries.
Scientific reports (2017-09-07)
Riyul Kim, Hyun Gyu Kim, Gisu Doo, Chanyong Choi, Soohyun Kim, Ju-Hyuk Lee, Jiyun Heo, Ho-Young Jung, Hee-Tak Kim
ABSTRACT
In this work, we present a 16 μm-thick Nafion-filled porous membrane for Zn/Br redox flow batteries (ZBBs). By using molecular dynamics simulation and dynamic light scattering analysis, we rationally design Nafion solution for Nafion impregnation into a porous polypropylene (PP) separator. A void-free Nafion/PP membrane is successfully fabricated by using NMP as a solvent for the Nafion solution. The resulting membrane shows a smaller area specific resistance in comparison with 600 μm-thick, commercial SF-600 porous membrane. Due to its dense morphology, Br
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)