MilliporeSigma
  • Synthesis, characterization, and antifungal evaluation of novel 1,2,3-triazolium-functionalized starch derivative.
Synthesis, characterization, and antifungal evaluation of novel 1,2,3-triazolium-functionalized starch derivative.
International journal of biological macromolecules (2017-04-05)
Wenqiang Tan, Jingjing Zhang, Fang Luan, Lijie Wei, Qing Li, Fang Dong, Zhanyong Guo
ABSTRACT
1,2,3-Triazolium-functionalized starch derivative was obtained by straightforward quaternization of the synthesized starch derivative bearing 1,2,3-triazole with benzyl bromide by combining the robust attributes of cuprous-catalyzed azide-alkyne cycloaddition. These novel starch derivatives were characterized by FTIR, UV-vis,
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
3-Butyn-1-ol, 97%
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%