MilliporeSigma
  • Inhibition of AMPK-related kinase 5 (ARK5) enhances cisplatin cytotoxicity in non-small cell lung cancer cells through regulation of epithelial-mesenchymal transition.
Inhibition of AMPK-related kinase 5 (ARK5) enhances cisplatin cytotoxicity in non-small cell lung cancer cells through regulation of epithelial-mesenchymal transition.
American journal of translational research (2017-05-05)
Minghui Li, Chengfei Zheng, Hongfei Xu, Wei He, Yongchun Ruan, Jianyong Ma, Junnan Zheng, Chengmeng Ye, Weidong Li
ABSTRACT
Lung cancer incidence and mortality rates are amongst the highest of all malignant tumors worldwide. ARK5 is a member of the human AMP-activated protein kinase (AMPK) family which is implicated in tumor survival and progression. The current study was designed to explore the role of ARK5 in resistance of non-small cell lung cancer (NSCLC) to cisplatin. We studied the sensitivity of two NSCLC cell lines, NCI-H1229 and A549, to cisplatin by using proliferation and cell viability assays. We then examined expression of ARK5, Twist, and the epithelial to mesenchymal transition (EMT) biomarkers, E-cadherin and Vimentin, by Western blot and immunofluorescence. We found that ARK5 downregulation significantly increased the cisplatin chemosensitivity of NSCLC cells, and that NCI-H1299 cells, which express high levels of ARK5 and possess a mesenchymal phenotype, were more resistant to cisplatin than A549 cells, which show low expression ARK5. Furthermore, siRNA-mediated silencing of ARK5 resulted in altered EMT patterns in NSCLC cells. These data support a role for ARK5 in regulating EMT in NSCLC cells. Together, our findings suggest that ARK5 is a potential drug target for combating drug resistance and regulating EMT in NSCLC cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human NUAK1