MilliporeSigma
  • miRNA-584-3p inhibits gastric cancer progression by repressing Yin Yang 1- facilitated MMP-14 expression.
miRNA-584-3p inhibits gastric cancer progression by repressing Yin Yang 1- facilitated MMP-14 expression.
Scientific reports (2017-08-23)
Liduan Zheng, Yajun Chen, Lin Ye, Wanju Jiao, Huajie Song, Hong Mei, Dan Li, Feng Yang, Huanhuan Li, Kai Huang, Qiangsong Tong
ABSTRACT
Recent evidence shows the emerging roles of promoter-targeting endogenous microRNAs (miRNAs) in regulating gene transcription. However, miRNAs affecting the transcription of matrix metalloproteinase 14 (MMP-14) in gastric cancer remain unknown. Herein, through integrative mining of public datasets, we identified the adjacent targeting sites of Yin Yang 1 (YY1) and miRNA-584-3p (miR-584-3p) within MMP-14 promoter. We demonstrated that YY1 directly targeted the MMP-14 promoter to facilitate its expression in gastric cancer cells. In contrast, miR-584-3p recognized its complementary site within MMP-14 promoter to suppress its expression. Mechanistically, miR-584-3p interacted with Argonaute 2 to recruit enhancer of zeste homolog 2 and euchromatic histone lysine methyltransferase 2, resulting in enrichment of repressive epigenetic markers and decreased binding of YY1 to MMP-14 promoter. miR-584-3p inhibited the in vitro and in vivo tumorigenesis and aggressiveness of gastric cancer cells through repressing YY1-facilitated MMP-14 expression. In clinical gastric cancer tissues, the expression of YY1 and miR-584-3p was positively or negatively correlated with MMP-14 levels. In addition, miR-584-3p and YY1 were independent prognostic factors associated with favorable and unfavorable outcome of gastric cancer patients, respectively. These data demonstrate that miR-584-3p directly targets the MMP-14 promoter to repress YY1-facilitated MMP-14 expression and inhibits the progression of gastric cancer.
Sigma-Aldrich
C646, ≥98% (HPLC)
Sigma-Aldrich
GSK343, ≥98% (HPLC)