MilliporeSigma
  • Electrical preconditioning of stem cells with a conductive polymer scaffold enhances stroke recovery.
Electrical preconditioning of stem cells with a conductive polymer scaffold enhances stroke recovery.
Biomaterials (2017-07-19)
Paul M George, Tonya M Bliss, Thuy Hua, Alex Lee, Byeongtaek Oh, Alexa Levinson, Swapnil Mehta, Guohua Sun, Gary K Steinberg
ABSTRACT
Exogenous human neural progenitor cells (hNPCs) are promising stroke therapeutics, but optimal delivery conditions and exact recovery mechanisms remain elusive. To further elucidate repair processes and improve stroke outcomes, we developed an electrically conductive, polymer scaffold for hNPC delivery. Electrical stimulation of hNPCs alters their transcriptome including changes to the VEGF-A pathway and genes involved in cell survival, inflammatory response, and synaptic remodeling. In our experiments, exogenous hNPCs were electrically stimulated (electrically preconditioned) via the scaffold 1 day prior to implantation. After in vitro stimulation, hNPCs on the scaffold are transplanted intracranially in a distal middle cerebral artery occlusion rat model. Electrically preconditioned hNPCs improved functional outcomes compared to unstimulated hNPCs or hNPCs where VEGF-A was blocked during in vitro electrical preconditioning. The ability to manipulate hNPCs via a conductive scaffold creates a new approach to optimize stem cell-based therapy and determine which factors (such as VEGF-A) are essential for stroke recovery.
Sigma-Aldrich
Anti-Nestin, Human Antibody, serum, from rabbit
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)