MilliporeSigma
  • Dynamic and thermodynamic mechanisms of TFA adsorption by particulate matter.
Dynamic and thermodynamic mechanisms of TFA adsorption by particulate matter.
Environmental pollution (Barking, Essex : 1987) (2017-04-04)
Junyu Guo, Zihan Zhai, Lei Wang, Ziyuan Wang, Jing Wu, Boya Zhang, Jianbo Zhang
ABSTRACT
Trifluoroacetic acid (TFA) in the atmosphere is produced by degradation of hydrochlorofluorocarbons and hydrofluorocarbons. In recent years, TFA has attracted global attention because of increased environmental concentrations, biological toxicity and accumulation in aqueous environments. This study focused on the mechanisms underlying the adsorption of TFA by particulate matter to identify the appropriate descriptive model for this process and thus improve estimation of TFA adsorption in future environmental monitoring. Onsite gas and particle phase sampling in Beijing, China, and subsequent measurement of TFA concentrations indicated that the TFA concentration in the gas phase (1396 ± 225 pg m
Sigma-Aldrich
2,4-Difluoroaniline, 99%