MilliporeSigma
  • Enantioselective analysis of ibuprofen enantiomers in mice plasma and tissues by high-performance liquid chromatography with fluorescence detection: Application to a pharmacokinetic study.
Enantioselective analysis of ibuprofen enantiomers in mice plasma and tissues by high-performance liquid chromatography with fluorescence detection: Application to a pharmacokinetic study.
Chirality (2017-07-06)
Katarzyna Przejczowska-Pomierny, Monika Włodyka, Agnieszka Cios, Elżbieta Wyska
ABSTRACT
A direct fluorometric high-performance liquid chromatography (HPLC) method was developed and validated for the analysis of ibuprofen enantiomers in mouse plasma (100 μl) and tissues (brain, liver, kidneys) using liquid-liquid extraction and 4-tertbutylphenoxyacetic acid as an internal standard. Separation of enantiomers was accomplished in a Chiracel OJ-H chiral column based on cellulose tris(4-methylbenzoate) coated on 5 μm silica-gel, 250 x 4.6 mm at 22 °C with a mobile phase composed of n-hexane, 2-propanol, and trifluoroacetic acid that were delivered in gradient elution at a flow rate of 1 ml min
Product Description
Supelco
Ibuprofen sodium salt, ≥98% (GC)