MilliporeSigma
  • Noncentrosomal microtubules regulate autophagosome transport through CAMSAP2-EB1 cross-talk.
Noncentrosomal microtubules regulate autophagosome transport through CAMSAP2-EB1 cross-talk.
FEBS letters (2017-07-21)
Jieli Wei, Honglin Xu, Wenxiang Meng
ABSTRACT
Microtubules (MTs) play essential roles in many steps of autophagy, an important degradation pathway in the maintenance of cellular homoeostasis. In many cells, MT networks are comprised of centrosomal MTs and noncentrosomal MTs. However, it is unknown whether noncentrosomal MTs and its binding proteins are involved in autophagy. Here, we show in HeLa cells that calmodulin-regulated spectrin-associated protein 2 (CAMSAP2), a noncentrosomal MT minus-end stabilizing protein, regulates retrograde transport of autophagosomes through MT dynamics. CAMSAP2 cooperates with EB1 to regulate end-binding protein 1 (EB1) behaviour at MT plus ends, MT growth directions and autophagosome transport. An association between CAMSAP2 and EB1 in the cytosol may modulate EB1 binding to MT plus ends. Collectively, our data indicate that noncentrosomal MTs regulate autophagy through a cross-talk between CAMSAP2 and EB1.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Tyrosine antibody produced in mouse, clone TUB-1A2, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Monoclonal Anti-Dynein (Intermediate Chain) antibody produced in mouse, clone 70.1, ascites fluid
Sigma-Aldrich
Anti-CAMSAP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit