MilliporeSigma
  • Curcumin and Salsalate Suppresses Colonic Inflammation and Procarcinogenic Signaling in High-Fat-Fed, Azoxymethane-Treated Mice.
Curcumin and Salsalate Suppresses Colonic Inflammation and Procarcinogenic Signaling in High-Fat-Fed, Azoxymethane-Treated Mice.
Journal of agricultural and food chemistry (2017-07-27)
Xian Wu, Anna C Pfalzer, Gar Yee Koh, Sanyuan Tang, Jimmy W Crott, Michael J Thomas, Mohsen Meydani, Joel B Mason
ABSTRACT
High-fat diets (HFDs) and excess adiposity increase proinflammatory cytokines in the colon, altering gene expression in a manner that promotes the development of colorectal cancer (CRC). Thus, compounds that reduce this biochemical inflammation are potential chemopreventive agents. Curcumin (CUR), a dietary polyphenol, and salsalate (SAL), a non-steroidal anti-inflammatory drug, are both anti-inflammatories. We investigated the inhibitory effects of CUR with or without SAL on inflammatory cytokines and procarcinogenic signaling in azoxymethane (AOM)-treated A/J mice. A sub-tumorigenic AOM dose was chosen to produce a biochemical and molecular procarcinogenic colonic environment without tumors. Mice were fed either a HFD (60% of kilocalories) or low-fat diet (LFD) (10% of kilocalories). One HFD treatment group received 0.2% CUR in the diet; one received 0.2% CUR + 0.15% SAL; and one received 0.4% CUR + 0.3% SAL. The HFD mice developed 30% greater fat mass than the LFD mice (p < 0.05). The colonic concentrations of interleukin-1β (IL-1β) and interleukin-6 (IL-6) in the HFD mice were decreased by 50-69% by the high-dose combination regimen (p < 0.015). Only the combination regimens significantly suppressed phosphorylation of protein kinase B (Akt) and nuclear factor-κB (NF-κB) p65 (p < 0.044). The combination of CUR and SAL reduces the concentration of proinflammatory cytokines and diminishes activation of Akt and NF-κB more effectively than CUR alone, providing a scientific basis for examining whether this combination mitigates the risk of CRC in obese individuals.
Sigma-Aldrich
Salsalate, ≥98% (HPLC)