MilliporeSigma
Charge transport mechanisms in sol-gel grown La
Physical chemistry chemical physics : PCCP (2017-02-01)
Eesh Vaghela, M J Keshvani, Keval Gadani, Zalak Joshi, Hetal Boricha, K Asokan, D Venkateshwarlu, V Ganesan, N A Shah, P S Solanki
ABSTRACT
In this communication, structural, microstructural, transport and magnetotransport properties are reported for La
Sigma-Aldrich
Manganese(II) acetate, 98%