MilliporeSigma
  • Inhibitor of H3K27 demethylase JMJD3/UTX GSK-J4 is a potential therapeutic option for castration resistant prostate cancer.
Inhibitor of H3K27 demethylase JMJD3/UTX GSK-J4 is a potential therapeutic option for castration resistant prostate cancer.
Oncotarget (2017-10-06)
Viacheslav M Morozov, Ying Li, Matthew M Clowers, Alexander M Ishov
ABSTRACT
Androgen receptor (AR) mediates initiation and progression of prostate cancer (PCa); AR-driven transcription is activated by binding of androgens to the ligand-binding domain (LBD) of AR. Androgen ablation therapy offers only a temporary relief of locally advanced and metastatic PCa, and the disease eventually recurs as a lethal castration-resistant PCa (CRPC) as there is no effective treatment for CRPC patients. Thus, it is critical to identify novel targeted and combinatorial regimens for clinical management of CRPC. Reduction of the repressive epigenetic modification H3K27me2/3 correlates with PCa aggressiveness, while corresponding demethylases JMJD3/UTX are overexpressed in PCa. We found that JMJD3/UTX inhibitor GSK-J4 reduced more efficiently proliferation of AR-ΔLBD cells (CRPC model) compared with isogenic AR-WT cells. Inhibition of JMJD3/UTX protects demethylation of H3K27Me2/3, thus reducing levels of H3k27Me1. We observed that the reduction dynamics of H3K27Me1 was faster and achieved at lower inhibitor concentrations in AR-ΔLBD cells, suggesting that inhibition of JMJD3/UTX diminished proliferation of these cells by hindering AR-driven transcription. In addition, we observed synergy between GSK-J4 and Cabazitaxel, a taxane derivative that is approved for CRPC treatment. Collectively, our results point at the H3K27 demethylation pathway as a new potential therapeutic target in CRPC patients.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
1 Estimated to ship on March 09, 2025
MYR 117.00
1 g
1 Estimated to ship on March 09, 2025
MYR 466.00
5 x 1 g
1 Estimated to ship on March 09, 2025
MYR 2,311.00
Sigma-Aldrich
GSK-J4, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
1 Estimated to ship on March 09, 2025
MYR 117.00
1 g
1 Estimated to ship on March 09, 2025
MYR 466.00
5 x 1 g
1 Estimated to ship on March 09, 2025
MYR 2,311.00
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
1 Estimated to ship on March 09, 2025
MYR 117.00
1 g
1 Estimated to ship on March 09, 2025
MYR 466.00
5 x 1 g
1 Estimated to ship on March 09, 2025
MYR 2,311.00
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
1 Estimated to ship on March 09, 2025
MYR 117.00
1 g
1 Estimated to ship on March 09, 2025
MYR 466.00
5 x 1 g
1 Estimated to ship on March 09, 2025
MYR 2,311.00