MilliporeSigma
  • Expression of sialyl-Tn sugar antigen in bladder cancer cells affects response to
Expression of sialyl-Tn sugar antigen in bladder cancer cells affects response to
Oncotarget (2017-09-15)
Paulo F Severino, Mariana Silva, Mylene Carrascal, Nadia Malagolini, Mariella Chiricolo, Giulia Venturi, Annalisa Astolfi, Mariangela Catera, Paula A Videira, Fabio Dall'Olio
ABSTRACT
The sialyl-Tn (sTn) antigen is an
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
MIBA, 96%