MilliporeSigma
  • Detection of antibiotics in chicken eggs obtained from supermarkets in Ho Chi Minh City, Vietnam.
Detection of antibiotics in chicken eggs obtained from supermarkets in Ho Chi Minh City, Vietnam.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2017-03-11)
Takahiro Yamaguchi, Masahiro Okihashi, Kazuo Harada, Yoshimasa Konishi, Kotaro Uchida, Mai Hoang Ngoc Do, Long Thi Bui, Thinh Duc Nguyen, Ha Bich Phan, Huong Dang Thien Bui, Phuc Do Nguyen, Keiji Kajimura, Yuko Kumeda, Chinh Van Dang, Kazumasa Hirata, Yoshimasa Yamamoto
ABSTRACT
The residual levels of antibiotics in Vietnamese eggs were monitored from 2014 to 2015. A total of 111 egg packages, distributed by 11 different companies, were collected from supermarkets in Ho Chi Minh City and the levels of 28 antibiotics were analyzed using liquid chromatography-tandem mass spectrometry (LC-MS/MS) screening method. Sixteen samples tested positive for antibiotics; a total of eight compounds (enrofloxacin, ciprofloxacin, norfloxacin, sulfadimethoxine, sulfamethazine, sulfamonomethoxine, tilmicosin and trimethoprim) were detected. Enrofloxacin was detected in eight samples, with two samples exhibiting concentrations exceeding 1,000 µg kg
Product Description
Supelco
Danofloxacin, VETRANAL®, analytical standard