MilliporeSigma
  • Increased expression of M1 and M2 phenotypic markers in isolated microglia after four-day binge alcohol exposure in male rats.
Increased expression of M1 and M2 phenotypic markers in isolated microglia after four-day binge alcohol exposure in male rats.
Alcohol (Fayetteville, N.Y.) (2017-08-02)
Hui Peng, Chelsea R Geil Nickell, Kevin Y Chen, Justin A McClain, Kimberly Nixon
ABSTRACT
Microglia activation and neuroinflammation are common features of neurodegenerative conditions, including alcohol use disorders (AUDs). When activated, microglia span a continuum of diverse phenotypes ranging from classically activated, pro-inflammatory (M1) microglia/macrophages to alternatively activated, growth-promoting (M2) microglia/macrophages. Identifying microglia phenotypes is critical for understanding the role of microglia in the pathogenesis of AUDs. Therefore, male rats were gavaged with 25% (w/v) ethanol or isocaloric control diet every 8 h for 4 days and sacrificed at 0, 2, 4, and 7 days after alcohol exposure (e.g., T0, T2, etc.). Microglia were isolated from hippocampus and entorhinal cortices by Percoll density gradient centrifugation. Cells were labeled with microglia surface antigens and analyzed by flow cytometry. Consistent with prior studies, isolated cells yielded a highly enriched population of brain macrophages/microglia (>95% pure), evidenced by staining for the macrophage/microglia antigen CD11b. Polarization states of CD11b
Product Description
Ethanol solution, NIST® SRM® 2899a, nominal 25% by mass