MilliporeSigma
  • Different Mutations Endowing Resistance to Acetyl-CoA Carboxylase Inhibitors Results in Changes in Ecological Fitness of
Different Mutations Endowing Resistance to Acetyl-CoA Carboxylase Inhibitors Results in Changes in Ecological Fitness of
Frontiers in plant science (2017-07-12)
Maor Matzrafi, Ofri Gerson, Baruch Rubin, Zvi Peleg
ABSTRACT
Various mutations altering the herbicide target site (TS), can lead to structural modifications that decrease binding efficiency and results in herbicide resistant weed. In most cases, such a mutation will be associated with ecological fitness penalty under herbicide free environmental conditions. Here we describe the effect of various mutations, endowing resistance to acetyl-CoA carboxylase (ACCase) inhibitors, on the ecological fitness penalty of
Sigma-Aldrich
1,2-Cyclohexanedione, 97%
Supelco
Pinoxaden, PESTANAL®, analytical standard