MilliporeSigma
  • Suppressing a Putative Sterol Carrier Gene Reduces Plasmodesmal Permeability and Activates Sucrose Transporter Genes during Cotton Fiber Elongation.
Suppressing a Putative Sterol Carrier Gene Reduces Plasmodesmal Permeability and Activates Sucrose Transporter Genes during Cotton Fiber Elongation.
The Plant cell (2017-07-28)
Zhiyuan Zhang, Yong-Ling Ruan, Na Zhou, Fang Wang, Xueying Guan, Lei Fang, Xiaoguang Shang, Wangzhen Guo, Shuijin Zhu, Tianzhen Zhang
ABSTRACT
Plasmodesmata (PDs) play vital roles in cell-to-cell communication and plant development. Emerging evidence suggests that sterols are involved in PD activity during cytokinesis. However, whether sterols contribute to PD gating between established cells remains unknown. Here, we isolated
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)