MilliporeSigma
  • Blood Biomarkers for the Early Diagnosis of Stroke: The Stroke-Chip Study.
Blood Biomarkers for the Early Diagnosis of Stroke: The Stroke-Chip Study.
Stroke (2017-07-19)
Alejandro Bustamante, Elena López-Cancio, Sara Pich, Anna Penalba, Dolors Giralt, Teresa García-Berrocoso, Carles Ferrer-Costa, Teresa Gasull, María Hernández-Pérez, Mónica Millan, Marta Rubiera, Pedro Cardona, Luis Cano, Helena Quesada, Mikel Terceño, Yolanda Silva, Mar Castellanos, Moisés Garces, Silvia Reverté, Xavier Ustrell, Rafael Marés, Joan Josep Baiges, Joaquín Serena, Francisco Rubio, Eduardo Salas, Antoni Dávalos, Joan Montaner
ABSTRACT
Stroke diagnosis could be challenging in the acute phase. We aimed to develop a blood-based diagnostic tool to differentiate between real strokes and stroke mimics and between ischemic and hemorrhagic strokes in the hyperacute phase. The Stroke-Chip was a prospective, observational, multicenter study, conducted at 6 Stroke Centers in Catalonia. Consecutive patients with suspected stroke were enrolled within the first 6 hours after symptom onset, and blood samples were drawn immediately after admission. A 21-biomarker panel selected among previous results and from the literature was measured by immunoassays. Outcomes were differentiation between real strokes and stroke mimics and between ischemic and hemorrhagic strokes. Predictive models were developed by combining biomarkers and clinical variables in logistic regression models. Accuracy was evaluated with receiver operating characteristic curves. From August 2012 to December 2013, 1308 patients were included (71.9% ischemic, 14.8% stroke mimics, and 13.3% hemorrhagic). For stroke versus stroke mimics comparison, no biomarker resulted included in the logistic regression model, but it was only integrated by clinical variables, with a predictive accuracy of 80.8%. For ischemic versus hemorrhagic strokes comparison, NT-proBNP (N-Terminal Pro-B-Type Natriuretic Peptide) >4.9 (odds ratio, 2.40; 95% confidence interval, 1.55-3.71; The studied biomarkers were not sufficient for an accurate differential diagnosis of stroke in the hyperacute setting. Additional discovery of new biomarkers and improvement on laboratory techniques seem necessary for achieving a molecular diagnosis of stroke.
Sigma-Aldrich
Human Serum, (from male AB clotted whole blood), USA origin, sterile-filtered