MilliporeSigma
  • Borane-catalysed postpolymerisation modification of the Si-H bonds in poly(phenylsilane).
Borane-catalysed postpolymerisation modification of the Si-H bonds in poly(phenylsilane).
Dalton transactions (Cambridge, England : 2003) (2017-04-01)
P T K Lee, L Rosenberg
ABSTRACT
B(C
Sigma-Aldrich
Allylbenzene, 98%
Sigma-Aldrich
2-Chloroethyl methyl ether, 98%
Sigma-Aldrich
Tris(pentafluorophenyl)borane, 95%