MilliporeSigma
  • (-)-Epigallocatechin-3-gallate alleviates doxorubicin-induced cardiotoxicity in sarcoma 180 tumor-bearing mice.
(-)-Epigallocatechin-3-gallate alleviates doxorubicin-induced cardiotoxicity in sarcoma 180 tumor-bearing mice.
Life sciences (2016-12-14)
Yu-Fei Yao, Xiang Liu, Wen-Juan Li, Zi-Wei Shi, Yu-Xin Yan, Le-Feng Wang, Ming Chen, Ming-Yong Xie
ABSTRACT
(-)-Epigallocatechin-3-gallate (EGCG), a major green tea polyphenol compound, plays an important role in the prevention of cardiovascular disease and cancer. The present study aimed to investigate the effects of EGCG on doxorubicin (DOX)-induced cardiotoxicity in Sarcoma 180 (S180) tumor-bearing mice. S180 tumor-bearing mice were established by subcutaneous inoculation of S180 cells attached to the axillary region. The extent of myocardial injury was accessed by the amount of lactate dehydrogenase (LDH) released in serum. Heart tissue was morphologically studied with transmission electron microscopy. Apoptosis, reactive oxygen species (ROS) generation, mitochondrial membrane potential (ΔѰm) as well as calcium concentration were measured by flow cytometric analysis. Expression levels of manganese superoxide dismutase (MnSOD) were analyzed by Western blot. Results showed that the combination with EGCG and DOX significantly inhibited tumor growth and enhanced induction of apoptosis compared with DOX alone. Moreover, administration of EGCG could suppress DOX-induced cardiotoxicity as evidenced by alleviating LDH release and apoptosis in cardiomyocyte. EGCG-evoked cardioprotection was in association with the increase of ΔѰm and MnSOD expression. EGCG was also found to attenuate ROS generation and myocardial calcium overload in Sarcoma 180 tumor-bearing mice subjected to DOX. EGCG alleviated DOX-induced cardiotoxicity possibly in part mediated by increasing of MnSOD and Ѱm, reducing myocardial calcium overload and subsequently attenuating the apoptosis and LDH release. Our findings suggest that co-administration of EGCG and DOX have potential as a feasible strategy to mitigate cardiotoxicity of DOX without compromising its chemotherapeutic value.
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate, BioReagent, suitable for fluorescence, ≥85% (HPCE)