MilliporeSigma
  • Running reorganizes the circuitry of one-week-old adult-born hippocampal neurons.
Running reorganizes the circuitry of one-week-old adult-born hippocampal neurons.
Scientific reports (2017-09-09)
Nirnath Sah, Benjamin D Peterson, Susan T Lubejko, Carmen Vivar, Henriette van Praag
ABSTRACT
Adult hippocampal neurogenesis is an important form of structural and functional plasticity in the mature mammalian brain. The existing consensus is that GABA regulates the initial integration of adult-born neurons, similar to neuronal development during embryogenesis. Surprisingly, virus-based anatomical tracing revealed that very young, one-week-old, new granule cells in male C57Bl/6 mice receive input not only from GABAergic interneurons, but also from multiple glutamatergic cell types, including mature dentate granule cells, area CA1-3 pyramidal cells and mossy cells. Consistently, patch-clamp recordings from retrovirally labeled new granule cells at 7-8 days post retroviral injection (dpi) show that these cells respond to NMDA application with tonic currents, and that both electrical and optogenetic stimulation can evoke NMDA-mediated synaptic responses. Furthermore, new dentate granule cell number, morphology and excitatory synaptic inputs at 7 dpi are modified by voluntary wheel running. Overall, glutamatergic and GABAergic innervation of newly born neurons in the adult hippocampus develops concurrently, and excitatory input is reorganized by exercise.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Picrotoxin, powder