MilliporeSigma
  • Critical roles of SMYD2-mediated β-catenin methylation for nuclear translocation and activation of Wnt signaling.
Critical roles of SMYD2-mediated β-catenin methylation for nuclear translocation and activation of Wnt signaling.
Oncotarget (2017-09-17)
Xiaolan Deng, Ryuji Hamamoto, Theodore Vougiouklakis, Rui Wang, Yuichiro Yoshioka, Takehiro Suzuki, Naoshi Dohmae, Yo Matsuo, Jae-Hyun Park, Yusuke Nakamura
ABSTRACT
Accumulation of β-catenin in the nucleus is a hallmark of activation of the Wnt/β-catenin signaling pathway, which drives development of a large proportion of human cancers. However, the mechanism of β-catenin nuclear translocation has not been well investigated. Here we report biological significance of SMYD2-mediated lysine 133 (K133) methylation of β-catenin on its nuclear translocation. Knockdown of SMYD2 attenuates the nuclear localization of β-catenin protein in human cancer cells. Consequently, transcriptional levels of well-known Wnt-signaling molecules,
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
LLY-507, ≥97% (HPLC)