MilliporeSigma
  • Efficacy of an Escherichia coli O157:H7 SRP Vaccine in Orally Challenged Goats and Strain Persistence Over Time.
Efficacy of an Escherichia coli O157:H7 SRP Vaccine in Orally Challenged Goats and Strain Persistence Over Time.
Foodborne pathogens and disease (2017-01-12)
Jacob M Swift, Derek M Foster, Anna T Rogers, Hannah J Sylvester, Emily H Griffith, Megan E Jacob
ABSTRACT
Small ruminants have been implicated in outbreaks of Escherichia coli O157:H7 at livestock exhibitions throughout the United States. Additionally, goat meat or milk may serve as a reservoir for foodborne transmission of the organism. These associations highlight the public health importance of an effective strategy to reduce E. coli O157:H7 shedding in goats. We examined the efficacy of the SRP
Product Description
Supelco
Florfenicol, analytical standard, for drug analysis