MilliporeSigma
  • Differential oxidation of protein-tyrosine phosphatases during zebrafish caudal fin regeneration.
Differential oxidation of protein-tyrosine phosphatases during zebrafish caudal fin regeneration.
Scientific reports (2017-08-18)
Wei Wu, Alexander James Hale, Simone Lemeer, Jeroen den Hertog
ABSTRACT
Zebrafish have the capacity to regenerate lost tissues and organs. Amputation of the caudal fin results in a rapid, transient increase in H
Sigma-Aldrich
Suberic acid bis(N-hydroxysuccinimide ester), ≥95%, powder