MilliporeSigma
  • The extracellular matrix protein agrin promotes heart regeneration in mice.
The extracellular matrix protein agrin promotes heart regeneration in mice.
Nature (2017-06-06)
Elad Bassat, Yara Eid Mutlak, Alex Genzelinakh, Ilya Y Shadrin, Kfir Baruch Umansky, Oren Yifa, David Kain, Dana Rajchman, John Leach, Daria Riabov Bassat, Yael Udi, Rachel Sarig, Irit Sagi, James F Martin, Nenad Bursac, Shenhav Cohen, Eldad Tzahor
ABSTRACT
The adult mammalian heart is non-regenerative owing to the post-mitotic nature of cardiomyocytes. The neonatal mouse heart can regenerate, but only during the first week of life. Here we show that changes in the composition of the extracellular matrix during this week can affect cardiomyocyte growth and differentiation in mice. We identify agrin, a component of neonatal extracellular matrix, as required for the full regenerative capacity of neonatal mouse hearts. In vitro, recombinant agrin promotes the division of cardiomyocytes that are derived from mouse and human induced pluripotent stem cells through a mechanism that involves the disassembly of the dystrophin-glycoprotein complex, and Yap- and ERK-mediated signalling. In vivo, a single administration of agrin promotes cardiac regeneration in adult mice after myocardial infarction, although the degree of cardiomyocyte proliferation observed in this model suggests that there are additional therapeutic mechanisms. Together, our results uncover a new inducer of mammalian heart regeneration and highlight fundamental roles of the extracellular matrix in cardiac repair.
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-MUSK antibody produced in mouse, clone 1F3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Anti-α-Dystroglycan Antibody, clone VIA4-1, culture supernatant, clone VIA4-1, Upstate®
Sigma-Aldrich
Anti-α-Actinin antibody produced in rabbit, whole antiserum