MilliporeSigma
Synthesis of coenzyme Q
Dalton transactions (Cambridge, England : 2003) (2017-04-01)
Olga V Zalomaeva, Vasilii Yu Evtushok, Gennadii M Maksimov, Raisa I Maksimovskaya, Oxana A Kholdeeva
ABSTRACT
The selective oxidation of methoxy/methyl-substituted arenes to the corresponding benzoquinones has been first realized using aqueous hydrogen peroxide as a green oxidant, acid tetrabutylammonium salts of the γ-Keggin divanadium-substituted phosphotungstate [γ-PW
Sigma-Aldrich
2,3-Dimethoxytoluene, 99%
Sigma-Aldrich
3,4,5-Trimethoxytoluene, 97%
Sigma-Aldrich
3,5-Dimethoxytoluene, 98%
Sigma-Aldrich
1,2,3-Trimethoxybenzene, 98%