MilliporeSigma
  • Sultamo-steroid analogues, Part VII: Synthesis of epi-androsterone derivatives with potential activity.
Sultamo-steroid analogues, Part VII: Synthesis of epi-androsterone derivatives with potential activity.
Archives of pharmacal research (1999-11-05)
S H Doss, N A Louca, G A Elmegeed, R M Mohareb
ABSTRACT
The reaction of epi-androsterone with cyanoacetyl hydrazine (2) gave the hydrazone derivative 3. The reactivity of 3 towards chemical reagents was studied to give pyridine, thiazole and thiophene derivatives.