MilliporeSigma
  • Clinical experience with silver sulfadiazine, a new topical agent for control of pseudomonas infections in burns
Clinical experience with silver sulfadiazine, a new topical agent for control of pseudomonas infections in burns
The Journal of Trauma and Acute Care Surgery (1969)
Stanford, W, et al.