MilliporeSigma
  • Recurrent De Novo and Biallelic Variation of ATAD3A, Encoding a Mitochondrial Membrane Protein, Results in Distinct Neurological Syndromes.
Recurrent De Novo and Biallelic Variation of ATAD3A, Encoding a Mitochondrial Membrane Protein, Results in Distinct Neurological Syndromes.
American journal of human genetics (2016-09-20)
Tamar Harel, Wan Hee Yoon, Caterina Garone, Shen Gu, Zeynep Coban-Akdemir, Mohammad K Eldomery, Jennifer E Posey, Shalini N Jhangiani, Jill A Rosenfeld, Megan T Cho, Stephanie Fox, Marjorie Withers, Stephanie M Brooks, Theodore Chiang, Lita Duraine, Serkan Erdin, Bo Yuan, Yunru Shao, Elie Moussallem, Costanza Lamperti, Maria A Donati, Joshua D Smith, Heather M McLaughlin, Christine M Eng, Magdalena Walkiewicz, Fan Xia, Tommaso Pippucci, Pamela Magini, Marco Seri, Massimo Zeviani, Michio Hirano, Jill V Hunter, Myriam Srour, Stefano Zanigni, Richard Alan Lewis, Donna M Muzny, Timothy E Lotze, Eric Boerwinkle, Richard A Gibbs, Scott E Hickey, Brett H Graham, Yaping Yang, Daniela Buhas, Donna M Martin, Lorraine Potocki, Claudio Graziano, Hugo J Bellen, James R Lupski
ABSTRACT
ATPase family AAA-domain containing protein 3A (ATAD3A) is a nuclear-encoded mitochondrial membrane protein implicated in mitochondrial dynamics, nucleoid organization, protein translation, cell growth, and cholesterol metabolism. We identified a recurrent de novo ATAD3A c.1582C>T (p.Arg528Trp) variant by whole-exome sequencing (WES) in five unrelated individuals with a core phenotype of global developmental delay, hypotonia, optic atrophy, axonal neuropathy, and hypertrophic cardiomyopathy. We also describe two families with biallelic variants in ATAD3A, including a homozygous variant in two siblings, and biallelic ATAD3A deletions mediated by nonallelic homologous recombination (NAHR) between ATAD3A and gene family members ATAD3B and ATAD3C. Tissue-specific overexpression of borR534W, the Drosophila mutation homologous to the human c.1582C>T (p.Arg528Trp) variant, resulted in a dramatic decrease in mitochondrial content, aberrant mitochondrial morphology, and increased autophagy. Homozygous null bor larvae showed a significant decrease of mitochondria, while overexpression of borWT resulted in larger, elongated mitochondria. Finally, fibroblasts of an affected individual exhibited increased mitophagy. We conclude that the p.Arg528Trp variant functions through a dominant-negative mechanism that results in small mitochondria that trigger mitophagy, resulting in a reduction in mitochondrial content. ATAD3A variation represents an additional link between mitochondrial dynamics and recognizable neurological syndromes, as seen with MFN2, OPA1, DNM1L, and STAT2 mutations.