MilliporeSigma
  • A new pyrazoline-based probe of quenched fluorescent reversible recognition for Cu
A new pyrazoline-based probe of quenched fluorescent reversible recognition for Cu
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-02-06)
Ying-Peng Zhang, Yu-Ying Dong, Yun-Shang Yang, Hui-Chen Guo, Bi-Xia Cao, Shi-Qi Sun
ABSTRACT
A new pyrazoline-based probe D was designed and synthesized, which can be used as a highly sensitive, selective and reversible recognizing fluorescent to detect Cu
Sigma-Aldrich
2-Hydroxyacetophenone, 98%
Sigma-Aldrich
9-Anthracenecarboxaldehyde, 97%