MilliporeSigma
  • Marmoset Cytochrome P450 3A4 Ortholog Expressed in Liver and Small-Intestine Tissues Efficiently Metabolizes Midazolam, Alprazolam, Nifedipine, and Testosterone.
Marmoset Cytochrome P450 3A4 Ortholog Expressed in Liver and Small-Intestine Tissues Efficiently Metabolizes Midazolam, Alprazolam, Nifedipine, and Testosterone.
Drug metabolism and disposition: the biological fate of chemicals (2017-02-16)
Shotaro Uehara, Yasuhiro Uno, Kazuyuki Nakanishi, Sakura Ishii, Takashi Inoue, Erika Sasaki, Hiroshi Yamazaki
ABSTRACT
Common marmosets (