MilliporeSigma
  • Zinc complexes of diflunisal: Synthesis, characterization, structure, antioxidant activity, and in vitro and in silico study of the interaction with DNA and albumins.
Zinc complexes of diflunisal: Synthesis, characterization, structure, antioxidant activity, and in vitro and in silico study of the interaction with DNA and albumins.
Journal of inorganic biochemistry (2017-02-24)
Alketa Tarushi, Chrisoula Kakoulidou, Catherine P Raptopoulou, Vassilis Psycharis, Dimitris P Kessissoglou, Ioanna Zoi, Athanasios N Papadopoulos, George Psomas
ABSTRACT
From the reaction of ZnCl
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Linoleic acid sodium salt, ≥97% (GC)