MilliporeSigma
  • Electrochemical DNA biosensor for detection of DNA damage induced by hydroxyl radicals.
Electrochemical DNA biosensor for detection of DNA damage induced by hydroxyl radicals.
Bioelectrochemistry (Amsterdam, Netherlands) (2017-03-18)
Andrea Hájková, Jiří Barek, Vlastimil Vyskočil
ABSTRACT
A simple electrochemical DNA biosensor based on a glassy carbon electrode (GCE) was prepared by adsorbing double-stranded DNA (dsDNA) onto the GCE surface and subsequently used for the detection of dsDNA damage induced by hydroxyl radicals. Investigation of the mutual interaction between hydroxyl radicals and dsDNA was conducted using a combination of several electrochemical detection techniques: square-wave voltammetry for direct monitoring the oxidation of dsDNA bases, and cyclic voltammetry and electrochemical impedance spectroscopy as indirect electrochemical methods making use of the redox-active indicator [Fe(CN)
Sigma-Aldrich
Deoxyribonucleic acid, low molecular weight from salmon sperm