MilliporeSigma
  • The potential of N-[2-(dimethylamino)ethyl]acridine-4-carboxamide to circumvent three multidrug-resistance phenotypes in vitro.
The potential of N-[2-(dimethylamino)ethyl]acridine-4-carboxamide to circumvent three multidrug-resistance phenotypes in vitro.
Cancer chemotherapy and pharmacology (1997-01-01)
R A Davey, G M Su, R M Hargrave, R M Harvie, B C Baguley, M W Davey
ABSTRACT
The effectiveness of N-[2-(dimethylamino)ethyl]acridine-4-carboxamide (DACA) relative to that of amsacrine, idarubicin, daunorubicin and paclitaxel against three different forms of multidrug resistance (MDR) was determined using two sublines of the CCRF-CEM human leukaemia cell line, the P-glyco-protein-expressing CEM/VLB100 subline and the MRP-expressing CEM/E1000 subline, and two extended-MDR sublines of the HL60 human leukaemia cell line, HL60/E8 and HL60/V8. DACA was effective against P-glycoprotein-mediated MDR and MRP-mediated MDR, whereas the extended-MDR phenotype showed only low levels of resistance (< 2-fold) to DACA. In comparison, idarubicin was ineffective against the MRP and extended-MDR phenotypes. Repeated exposure of the K562 human leukaemia cell line to DACA (55, 546 or 1092 nM for 3 days over 10 weeks) did not result in the development of any significant drug resistance. We conclude that DACA has the potential to treat refractory leukaemia.
Sigma-Aldrich
Amsacrine hydrochloride, ≥98% (TLC), powder