MilliporeSigma
  • Synthesis, structure elucidation and biological evaluation of triple bridged dinuclear copper(II) complexes as anticancer and antioxidant/anti-inflammatory agents.
Synthesis, structure elucidation and biological evaluation of triple bridged dinuclear copper(II) complexes as anticancer and antioxidant/anti-inflammatory agents.
Materials science & engineering. C, Materials for biological applications (2017-05-10)
Z Boulsourani, S Katsamakas, G D Geromichalos, V Psycharis, C P Raptopoulou, D Hadjipavlou-Litina, E Yiannaki, C Dendrinou-Samara
ABSTRACT
Seeking for copper based metallo-therapeutics, three triple bridged dinuclear copper(II) complexes [Cu
Sigma-Aldrich
2-Thiopheneacetic acid, 98%