MilliporeSigma
  • Cobll1 is linked to drug resistance and blastic transformation in chronic myeloid leukemia.
Cobll1 is linked to drug resistance and blastic transformation in chronic myeloid leukemia.
Leukemia (2017-02-25)
S H Han, S-H Kim, H-J Kim, Y Lee, S-Y Choi, G Park, D-H Kim, A Lee, J Kim, J-M Choi, Y Kim, K Myung, H Kim, D-W Kim
ABSTRACT
Drug resistance to BCR-ABL1 tyrosine kinase inhibitor (TKI) and disease progression to blast crisis (BC) are major clinical problems in chronic myeloid leukemia (CML); however, underlying mechanisms governing this process remain to be elucidated. Here, we report Cordon-bleu protein-like 1 (Cobll1) as a distinct molecular marker associated with drug resistance as well as progression to BC. In detail, Cobll1 increases IKKγ stability, leading to NF-κB activation and reduction of nilotinib-dependent apoptosis, suggesting Cobll1-mediated NF-κB could be involved in drug resistance. Recently, NF-κB signalling has been highlighted as a core mechanism for chronic phase (CP)-BC progression, stem cell survival and tyrosine kinase inhibitor resistance. We also demonstrated that high expression of Cobll1 confers drug resistance to tyrosine kinase inhibitors in CML cell line as well as patient samples. The analysis of large sets of primary CML samples (n=90) shows that Cobll1 expression is dramatically increased in BC but not in CP, which is correlated with a poor survival rate (P=0.002). Moreover, our studies show that Cobll1 is highly expressed in CD34
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Nilotinib