MilliporeSigma
  • Successful newborn screening for Gaucher disease using fluorometric assay in China.
Successful newborn screening for Gaucher disease using fluorometric assay in China.
Journal of human genetics (2017-03-31)
Lulu Kang, Xia Zhan, Xuefan Gu, Huiwen Zhang
ABSTRACT
Gaucher disease (GD) is an inherited metabolic disorder that involves accumulation of glycolipid glucocerebroside in monocyte-macrophage cells, which can result in multiple organ damage. Enzyme replacement and substrate reduction therapies have improved the potential for early diagnosis and treatment. Determining the true incidence of this rare disease is critical for relevant policy establishment. Newborn screening allows for early diagnosis and an comparatively accurate incidence of GD. A fluorometric method to detect acid β-glucocerebrosidase (GBA) activity on a dried blood spot punch was developed. Validity and feasibility of the fluorometric method was demonstrated by examining 116 healthy controls, 19 confirmed GD patients and 19 obligate carriers. GBA activity was measured on dried blood spots of 80 855 newborns. Samples from positively screened newborns were reanalyzed by a leukocyte GBA activity test and GBA gene analysis. Plasma glucosylsphingosine level was determined as a biomarker of the pathophysiology of GD. GD patients were distinguished from healthy controls and obligate carriers using the fluorometric method. Mean GBA activity in newborn screening specimens was 145.69±44.76 μmol l
Sigma-Aldrich
Glucosylsphingosine, ≥98.0% (TLC)