MilliporeSigma
  • Overexpression of KCNJ2 in induced pluripotent stem cell-derived cardiomyocytes for the assessment of QT-prolonging drugs.
Overexpression of KCNJ2 in induced pluripotent stem cell-derived cardiomyocytes for the assessment of QT-prolonging drugs.
Journal of pharmacological sciences (2017-06-16)
Min Li, Yasunari Kanda, Takashi Ashihara, Tetsuo Sasano, Yuji Nakai, Masami Kodama, Erina Hayashi, Yuko Sekino, Tetsushi Furukawa, Junko Kurokawa
ABSTRACT
Human induced pluripotent stem cell (hiPSC)-derived cardiomyocytes hold great potentials to predict pro-arrhythmic risks in preclinical cardiac safety screening, although the hiPSC cardiomyocytes exhibit rather immature functional and structural characteristics, including spontaneous activity. Our physiological characterization and mathematical simulation showed that low expression of the inward-rectifier potassium (I
Sigma-Aldrich
E-4031, ≥98% (HPLC), lyophilized powder