MilliporeSigma
  • Poplar MYB115 and MYB134 Transcription Factors Regulate Proanthocyanidin Synthesis and Structure.
Poplar MYB115 and MYB134 Transcription Factors Regulate Proanthocyanidin Synthesis and Structure.
Plant physiology (2017-03-30)
Amy Midori James, Dawei Ma, Robin Mellway, Andreas Gesell, Kazuko Yoshida, Vincent Walker, Lan Tran, Don Stewart, Michael Reichelt, Jussi Suvanto, Juha-Pekka Salminen, Jonathan Gershenzon, Armand Séguin, C Peter Constabel
ABSTRACT
The accumulation of proanthocyanidins is regulated by a complex of transcription factors composed of R2R3 MYB, basic helix-loop-helix, and WD40 proteins that activate the promoters of biosynthetic genes. In poplar (genus
Sigma-Aldrich
Indole-3-butyric acid potassium salt, BioReagent, suitable for plant cell culture