MilliporeSigma
  • Rapid determination of 9 aromatic amines in mainstream cigarette smoke by modified dispersive liquid liquid microextraction and ultraperformance convergence chromatography tandem mass spectrometry.
Rapid determination of 9 aromatic amines in mainstream cigarette smoke by modified dispersive liquid liquid microextraction and ultraperformance convergence chromatography tandem mass spectrometry.
Journal of chromatography. A (2017-06-07)
Huimin Deng, Fei Yang, Zhonghao Li, Zhaoyang Bian, Ziyan Fan, Ying Wang, Shanshan Liu, Gangling Tang
ABSTRACT
Aromatic amines in mainstream cigarette smoke have long been monitored due to their carcinogenic toxicity. In this work, a reliable and rapid method was developed for the simultaneous determination of 9 aromatic amines in mainstream cigarette smoke by modified dispersive liquid liquid microextraction (DLLME) and ultraperformance convergence chromatography tandem mass spectrometry (UPC
Sigma-Aldrich
3-Aminobiphenyl, 97%