MilliporeSigma
  • Sensitization, energy transfer and infra-red emission decay modulation in Yb
Sensitization, energy transfer and infra-red emission decay modulation in Yb
Scientific reports (2017-07-13)
Haizhou Lu, Yu Peng, Huanqing Ye, Xianjin Cui, Jianxu Hu, Hang Gu, Andrei N Khlobystov, Mark A Green, Philip J Blower, Peter B Wyatt, William P Gillin, Ignacio Hernández
ABSTRACT
Infra-red emission (980 nm) of sub 10 nm Yb
Sigma-Aldrich
1-Octadecene, technical grade, 90%