MilliporeSigma
  • Activity-Dependent Sulfhydration Signal Controls N-Methyl-D-Aspartate Subtype Glutamate Receptor-Dependent Synaptic Plasticity via Increasing d-Serine Availability.
Activity-Dependent Sulfhydration Signal Controls N-Methyl-D-Aspartate Subtype Glutamate Receptor-Dependent Synaptic Plasticity via Increasing d-Serine Availability.
Antioxidants & redox signaling (2017-01-05)
Yuan-Long Li, Peng-Fei Wu, Jian-Guo Chen, Sheng Wang, Qian-Qian Han, Dan Li, Wen Wang, Xin-Lei Guan, Di Li, Li-Hong Long, Jian-Geng Huang, Fang Wang
ABSTRACT
Reactive sulfur species, including hydrogen sulfide (H In this study, we found that increased synaptic activity triggered sulfide generation and protein sulfhydration. Activity-triggered sulfide production was essential for N-methyl-D-aspartate subtype glutamate receptor (NMDAR)-dependent LTP via maintaining the availability of d-serine, a primary coagonist for synaptic NMDARs. Genetic knockdown of cystathionine β-synthase, not cystathionine γ-lyase, impaired LTP. H
Sigma-Aldrich
D-Amino Acid Oxidase from porcine kidney, powder, ≥1.5 units/mg solid
Millipore
Streptavidin−Agarose from Streptomyces avidinii, buffered aqueous suspension