MilliporeSigma
  • Methane production in an anaerobic osmotic membrane bioreactor using forward osmosis: Effect of reverse salt flux.
Methane production in an anaerobic osmotic membrane bioreactor using forward osmosis: Effect of reverse salt flux.
Bioresource technology (2017-05-23)
Sheng Li, Youngjin Kim, Sherub Phuntsho, Laura Chekli, Ho Kyong Shon, TorOve Leiknes, Noreddine Ghaffour
ABSTRACT
This study investigated the impact of reverse salt flux (RSF) on microbe community and bio-methane production in a simulated fertilizer driven FO-AnMBR system using KCl, KNO