MilliporeSigma
  • nr3c1 null mutant zebrafish are viable and reveal DNA-binding-independent activities of the glucocorticoid receptor.
nr3c1 null mutant zebrafish are viable and reveal DNA-binding-independent activities of the glucocorticoid receptor.
Scientific reports (2017-07-01)
N Facchinello, T Skobo, G Meneghetti, E Colletti, A Dinarello, N Tiso, R Costa, G Gioacchini, O Carnevali, F Argenton, L Colombo, L Dalla Valle
ABSTRACT
Glucocorticoids (GCs) play important roles in developmental and physiological processes through the transcriptional activity of their cognate receptor (Gr). Using CRISPR/Cas9 technology, we established a zebrafish null Gr mutant line and compared its phenotypes with wild type and a zebrafish line with partially silenced gr (gr
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Dextran sulfate sodium salt, Mr ~40,000
Sigma-Aldrich
Triton X-100, laboratory grade