MilliporeSigma
  • Structural Characterization of Neutral Saccharides by Negative Ion MALDI Mass Spectrometry Using a Superbasic Proton Sponge as Deprotonating Matrix.
Structural Characterization of Neutral Saccharides by Negative Ion MALDI Mass Spectrometry Using a Superbasic Proton Sponge as Deprotonating Matrix.
Journal of the American Society for Mass Spectrometry (2017-05-04)
Cosima Damiana Calvano, Tommaso R I Cataldi, Julius F Kögel, Antonio Monopoli, Francesco Palmisano, Jorge Sundermeyer
ABSTRACT
The superbasic proton sponge 1,8-bis(tripyrrolidinylphosphazenyl)naphthalene (TPPN) has been successfully employed for the structural characterization of neutral saccharides, cyclodextrins, and saccharide alditols by matrix assisted laser desorption/ionization tandem mass spectrometry (MALDI-MS/MS). Owing to its inherently high basicity, TPPN is capable of deprotonating neutral carbohydrates (M) providing an efficient and simple way to produce gas-phase [M - H]
Sigma-Aldrich
β-Gentiobiose, ≥85% (remainder primarily α-anomer)