MilliporeSigma
  • Impaired hippocampal glucose metabolism during and after flurothyl-induced seizures in mice: Reduced phosphorylation coincides with reduced activity of pyruvate dehydrogenase.
Impaired hippocampal glucose metabolism during and after flurothyl-induced seizures in mice: Reduced phosphorylation coincides with reduced activity of pyruvate dehydrogenase.
Epilepsia (2017-06-21)
Tanya S McDonald, Karin Borges
ABSTRACT
To determine changes in glucose metabolism and the enzymes involved in the hippocampus ictally and postictally in the acute mouse flurothyl seizure model. [U- During seizures, total lactate levels increased 1.7-fold; however, [M + 3] enrichment of both lactate and alanine were reduced by 30% and 43%, respectively, along with a 28% decrease in phosphofructokinase activity. Postictally the % Here, we show that the increase of lactate levels during flurothyl seizures is from a source other than [U-
Sigma-Aldrich
Succinic acid-2,2,3,3-d4, 98 atom % D