MilliporeSigma
  • Recent increases in terrestrial carbon uptake at little cost to the water cycle.
Recent increases in terrestrial carbon uptake at little cost to the water cycle.
Nature communications (2017-07-26)
Lei Cheng, Lu Zhang, Ying-Ping Wang, Josep G Canadell, Francis H S Chiew, Jason Beringer, Longhui Li, Diego G Miralles, Shilong Piao, Yongqiang Zhang
ABSTRACT
Quantifying the responses of the coupled carbon and water cycles to current global warming and rising atmospheric CO
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)